RIE240870574 Rieke Metals, Inc.

2-Methyl-5-(3-nitrobenzoyl)pyridine

  • MDL number MFCD13153229
  • Molecular Weight : 242.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5623-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5623-30
Availability details for
Quantity
  • Availability
  • Add to Cart