RIE240870579 Rieke Metals, Inc.

5-(4-Ethylbenzoyl)-2-methylpyridine

  • MDL number MFCD13153234
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-ethylphenyl)(6-methyl-3-pyridinyl)methanone
Inchi Code 1S/C15H15NO/c1-3-12-5-8-13(9-6-12)15(17)14-7-4-11(2)16-10-14/h4-10H,3H2,1-2H3
Inchi Key RGIVPUMPGGLKLH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5623-35a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5623-35a
Availability details for
Quantity
  • Availability
  • Add to Cart