RIE240870587 Rieke Metals, Inc.

5-(4-Butoxybenzoyl)-2-methylpyridine

  • MDL number MFCD13153242
  • Molecular Weight : 269.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-butoxyphenyl)(6-methyl-3-pyridinyl)methanone
Inchi Code 1S/C17H19NO2/c1-3-4-11-20-16-9-7-14(8-10-16)17(19)15-6-5-13(2)18-12-15/h5-10,12H,3-4,11H2,1-2H3
Inchi Key MHPQCUORULUBBI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5623-36c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5623-36c
Availability details for
Quantity
  • Availability
  • Add to Cart