RIE240870590 Rieke Metals, Inc.

5-(4-Heptyloxybenzoyl)-2-methylpyridine

  • MDL number MFCD13153245
  • Molecular Weight : 311.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5623-36F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5623-36F
Availability details for
Quantity
  • Availability
  • Add to Cart