RIE240870594 Rieke Metals, Inc.

5-(2,6-Difluorobenzoyl)-2-methylpyridine

  • MDL number MFCD13153249
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5623-40

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5623-40
Availability details for
Quantity
  • Availability
  • Add to Cart