RIE240870596 Rieke Metals, Inc.

5-(3,5-Difluorobenzoyl)-2-methylpyridine

  • MDL number MFCD13153251
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-difluorophenyl)(6-methyl-3-pyridinyl)methanone
Inchi Code 1S/C13H9F2NO/c1-8-2-3-9(7-16-8)13(17)10-4-11(14)6-12(15)5-10/h2-7H,1H3
Inchi Key ZMVQQMODGIKUBA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5623-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5623-42
Availability details for
Quantity
  • Availability
  • Add to Cart