RIE240870600 Rieke Metals, Inc.

5-(2,6-Dichlorobenzoyl)-2-methylpyridine

  • MDL number MFCD13153255
  • Molecular Weight : 266.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5623-46

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5623-46
Availability details for
Quantity
  • Availability
  • Add to Cart