RIE240870605 Rieke Metals, Inc.

5-(2,5-Dimethoxybenzoyl)-2-methylpyridine

  • MDL number MFCD13153260
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,5-dimethoxyphenyl)(6-methyl-3-pyridinyl)methanone
Inchi Code 1S/C15H15NO3/c1-10-4-5-11(9-16-10)15(17)13-8-12(18-2)6-7-14(13)19-3/h4-9H,1-3H3
Inchi Key CJDSUPZPCLJBRR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5623-51

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5623-51
Availability details for
Quantity
  • Availability
  • Add to Cart