RIE240870613 Rieke Metals, Inc.

5-(3,4-Dimethylbenzoyl)-2-methylpyridine

  • MDL number MFCD13153268
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-dimethylphenyl)(6-methyl-3-pyridinyl)methanone
Inchi Code 1S/C15H15NO/c1-10-4-6-13(8-11(10)2)15(17)14-7-5-12(3)16-9-14/h4-9H,1-3H3
Inchi Key AIAOAJJUOQPRRF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5623-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5623-59
Availability details for
Quantity
  • Availability
  • Add to Cart