RIE240870621 Rieke Metals, Inc.

4-(2-Methylbenzoyl)-2-methylpyridine

  • MDL number MFCD13153276
  • Molecular Weight : 211.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5624-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-05
Availability details for
Quantity
  • Availability
  • Add to Cart