RIE240870627 Rieke Metals, Inc.

4-(2-Fluorobenzoyl)-2-methylpyridine

  • MDL number MFCD13153282
  • Molecular Weight : 215.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5624-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-11
Availability details for
Quantity
  • Availability
  • Add to Cart