RIE240870633 Rieke Metals, Inc.

4-(2-Bromobenzoyl)-2-methylpyridine

  • MDL number MFCD13153288
  • Molecular Weight : 276.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-bromophenyl)(2-methyl-4-pyridinyl)methanone
Inchi Code 1S/C13H10BrNO/c1-9-8-10(6-7-15-9)13(16)11-4-2-3-5-12(11)14/h2-8H,1H3
Inchi Key OQGONLRJKKQBII-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5624-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-17
Availability details for
Quantity
  • Availability
  • Add to Cart