RIE240870636 Rieke Metals, Inc.

4-(2-Iodobenzoyl)-2-methylpyridine

  • MDL number MFCD13153291
  • Molecular Weight : 323.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-iodophenyl)(2-methyl-4-pyridinyl)methanone
Inchi Code 1S/C13H10INO/c1-9-8-10(6-7-15-9)13(16)11-4-2-3-5-12(11)14/h2-8H,1H3
Inchi Key AUPINVBCUPGVDW-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5624-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-20
Availability details for
Quantity
  • Availability
  • Add to Cart