RIE240870637 Rieke Metals, Inc.

4-(3-Iodobenzoyl)-2-methylpyridine

  • MDL number MFCD13153292
  • Molecular Weight : 323.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-iodophenyl)(2-methyl-4-pyridinyl)methanone
Inchi Code 1S/C13H10INO/c1-9-7-11(5-6-15-9)13(16)10-3-2-4-12(14)8-10/h2-8H,1H3
Inchi Key WIINDMGMPGKMMB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5624-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-21
Availability details for
Quantity
  • Availability
  • Add to Cart