RIE240870640 Rieke Metals, Inc.

4-(4-Cyanobenzoyl)-2-methylpyridine

  • MDL number MFCD13153295
  • Molecular Weight : 222.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2-methylisonicotinoyl)benzonitrile
Inchi Code 1S/C14H10N2O/c1-10-8-13(6-7-16-10)14(17)12-4-2-11(9-15)3-5-12/h2-8H,1H3
Inchi Key MSBGLMFSXKALPS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5624-25

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-25
Availability details for
Quantity
  • Availability
  • Add to Cart