RIE240870642 Rieke Metals, Inc.

2-Methyl-4-(4-phenoxybenzoyl)pyridine

  • MDL number MFCD13153297
  • Molecular Weight : 289.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methyl-4-pyridinyl)(4-phenoxyphenyl)methanone
Inchi Code 1S/C19H15NO2/c1-14-13-16(11-12-20-14)19(21)15-7-9-18(10-8-15)22-17-5-3-2-4-6-17/h2-13H,1H3
Inchi Key OPQRTOGFBMNHSN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5624-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-28
Availability details for
Quantity
  • Availability
  • Add to Cart