RIE240870647 Rieke Metals, Inc.

4-(4-tert-Butylbenzoyl)-2-methylpyridine

  • MDL number MFCD13153302
  • Molecular Weight : 253.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5624-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-34
Availability details for
Quantity
  • Availability
  • Add to Cart