RIE240870649 Rieke Metals, Inc.

2-Methyl-4-(4-propylbenzoyl)pyridine

  • MDL number MFCD13153304
  • Molecular Weight : 239.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5624-35B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-35B
Availability details for
Quantity
  • Availability
  • Add to Cart