RIE240870656 Rieke Metals, Inc.

4-(4-Butoxybenzoyl)-2-methylpyridine

  • MDL number MFCD13153311
  • Molecular Weight : 269.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-butoxyphenyl)(2-methyl-4-pyridinyl)methanone
Inchi Code 1S/C17H19NO2/c1-3-4-11-20-16-7-5-14(6-8-16)17(19)15-9-10-18-13(2)12-15/h5-10,12H,3-4,11H2,1-2H3
Inchi Key NQASOZJXGWMGOM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5624-36c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-36c
Availability details for
Quantity
  • Availability
  • Add to Cart