RIE240870660 Rieke Metals, Inc.

4-(2,3-Difluorobenzoyl)-2-methylpyridine

  • MDL number MFCD13153315
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5624-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-37
Availability details for
Quantity
  • Availability
  • Add to Cart