RIE240870661 Rieke Metals, Inc.

4-(2,4-Difluorobenzoyl)-2-methylpyridine

  • MDL number MFCD13153316
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,4-difluorophenyl)(2-methyl-4-pyridinyl)methanone
Inchi Code 1S/C13H9F2NO/c1-8-6-9(4-5-16-8)13(17)11-3-2-10(14)7-12(11)15/h2-7H,1H3
Inchi Key SMWCIMMATRWERL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5624-38

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-38
Availability details for
Quantity
  • Availability
  • Add to Cart