RIE240870664 Rieke Metals, Inc.

4-(3,4-Difluorobenzoyl)-2-methylpyridine

  • MDL number MFCD13153319
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5624-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-41
Availability details for
Quantity
  • Availability
  • Add to Cart