RIE240870668 Rieke Metals, Inc.

4-(2,5-Dichlorobenzoyl)-2-methylpyridine

  • MDL number MFCD13153323
  • Molecular Weight : 266.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,5-dichlorophenyl)(2-methyl-4-pyridinyl)methanone
Inchi Code 1S/C13H9Cl2NO/c1-8-6-9(4-5-16-8)13(17)11-7-10(14)2-3-12(11)15/h2-7H,1H3
Inchi Key MCJHJGCHQSPWJI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5624-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-45
Availability details for
Quantity
  • Availability
  • Add to Cart