RIE240870671 Rieke Metals, Inc.

4-(3,5-Dichlorobenzoyl)-2-methylpyridine

  • MDL number MFCD13153326
  • Molecular Weight : 266.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-dichlorophenyl)(2-methyl-4-pyridinyl)methanone
Inchi Code 1S/C13H9Cl2NO/c1-8-4-9(2-3-16-8)13(17)10-5-11(14)7-12(15)6-10/h2-7H,1H3
Inchi Key RJMAUHYBZZUBLC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5624-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-48
Availability details for
Quantity
  • Availability
  • Add to Cart