RIE240870673 Rieke Metals, Inc.

4-(2,4-Dimethoxybenzoyl)-2-methylpyridine

  • MDL number MFCD13153328
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,4-dimethoxyphenyl)(2-methyl-4-pyridinyl)methanone
Inchi Code 1S/C15H15NO3/c1-10-8-11(6-7-16-10)15(17)13-5-4-12(18-2)9-14(13)19-3/h4-9H,1-3H3
Inchi Key ZHMIWUIXGQANDY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5624-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-50
Availability details for
Quantity
  • Availability
  • Add to Cart