RIE240870680 Rieke Metals, Inc.

4-(2,5-Dimethylbenzoyl)-2-methylpyridine

  • MDL number MFCD13153335
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,5-dimethylphenyl)(2-methyl-4-pyridinyl)methanone
Inchi Code 1S/C15H15NO/c1-10-4-5-11(2)14(8-10)15(17)13-6-7-16-12(3)9-13/h4-9H,1-3H3
Inchi Key YCAMZZCIWITRGB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5624-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5624-57
Availability details for
Quantity
  • Availability
  • Add to Cart