RIE240870697 Rieke Metals, Inc.

2-(4-Chlorophenyl)pyridine

  • MDL number MFCD06201380
  • Molecular Weight : 189.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/11042

Safety Information

MSDS http://riekemetals.com/products-services/catnum/11042
Availability details for
Quantity
  • Availability
  • Add to Cart