RIE240870699 Rieke Metals, Inc.

4-(2-Pyridyl)aniline

  • MDL number MFCD04116305
  • Cas Number : 18471-73-3
  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/11500

Safety Information

MSDS http://riekemetals.com/products-services/catnum/11500
Availability details for
Quantity
  • Availability
  • Add to Cart