RIE240870700 Rieke Metals, Inc.

2-(4-Fluorobenzoyl)-3-methylpyridine

  • MDL number MFCD13152778
  • Molecular Weight : 215.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-fluorophenyl)(3-methyl-2-pyridinyl)methanone
Inchi Code 1S/C13H10FNO/c1-9-3-2-8-15-12(9)13(16)10-4-6-11(14)7-5-10/h2-8H,1H3
Inchi Key APUVRZPRUXFUAX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5615-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-13
Availability details for
Quantity
  • Availability
  • Add to Cart