RIE240870740 Rieke Metals, Inc.

3-Benzoylquinoline

  • MDL number MFCD12877737
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5621-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-01
Availability details for
Quantity
  • Availability
  • Add to Cart