RIE240870742 Rieke Metals, Inc.

3-(3-Methoxybenzoyl)quinoline

  • MDL number MFCD13153078
  • Molecular Weight : 263.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-methoxyphenyl)(3-quinolinyl)methanone
Inchi Code 1S/C17H13NO2/c1-20-15-7-4-6-13(10-15)17(19)14-9-12-5-2-3-8-16(12)18-11-14/h2-11H,1H3
Inchi Key MRGSZDQXVMCPNR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5621-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-03
Availability details for
Quantity
  • Availability
  • Add to Cart