RIE240870745 Rieke Metals, Inc.

4-(3-Methylbenzoyl)quinoline

  • MDL number MFCD12833805
  • Molecular Weight : 247.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5621-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-06
Availability details for
Quantity
  • Availability
  • Add to Cart