RIE240870747 Rieke Metals, Inc.

2-Benzoyl-4-methylpyridine

  • MDL number MFCD00666136
  • Molecular Weight : 197.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-methyl-2-pyridinyl)(phenyl)methanone
Inchi Code 1S/C13H11NO/c1-10-7-8-14-12(9-10)13(15)11-5-3-2-4-6-11/h2-9H,1H3
Inchi Key UBHIEEUGUQXEBO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5614-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-01
Availability details for
Quantity
  • Availability
  • Add to Cart