RIE240870757 Rieke Metals, Inc.

4-(3-Bromobenzoyl)quinoline

  • MDL number MFCD12877736
  • Molecular Weight : 312.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5621-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-18
Availability details for
Quantity
  • Availability
  • Add to Cart