RIE240870761 Rieke Metals, Inc.

4-Methoxy-3'-trifluoromethylbenzophenone

  • MDL number MFCD13152564
  • Molecular Weight : 280.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5051-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5051-44
Availability details for
Quantity
  • Availability
  • Add to Cart