RIE240870787 Rieke Metals, Inc.

3-(3,4-Difluorobenzoyl)quinoline

  • MDL number MFCD13153114
  • Molecular Weight : 269.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-difluorophenyl)(3-quinolinyl)methanone
Inchi Code 1S/C16H9F2NO/c17-13-6-5-11(8-14(13)18)16(20)12-7-10-3-1-2-4-15(10)19-9-12/h1-9H
Inchi Key UFNCLQVQTCTEIJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5621-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5621-41
Availability details for
Quantity
  • Availability
  • Add to Cart