RIE240870792 Rieke Metals, Inc.

2-(4-Fluorobenzoyl)-6-methylpyridine

  • MDL number MFCD13152578
  • Molecular Weight : 215.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5612-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5612-13
Availability details for
Quantity
  • Availability
  • Add to Cart