RIE240870968 Rieke Metals, Inc.

2-Chloro-5-(4-methylbenzoyl)pyridine

  • MDL number MFCD13153343
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5625-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-07
Availability details for
Quantity
  • Availability
  • Add to Cart