RIE240870975 Rieke Metals, Inc.

5-(2-Chlorobenzoyl)-2-chloropyridine

  • MDL number MFCD13153349
  • Molecular Weight : 252.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chlorophenyl)(6-chloro-3-pyridinyl)methanone
Inchi Code 1S/C12H7Cl2NO/c13-10-4-2-1-3-9(10)12(16)8-5-6-11(14)15-7-8/h1-7H
Inchi Key SQSYIAYULMJWAD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5625-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-14
Availability details for
Quantity
  • Availability
  • Add to Cart