RIE240870976 Rieke Metals, Inc.

5-(3-Chlorobenzoyl)-2-chloropyridine

  • MDL number MFCD07775838
  • Molecular Weight : 252.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chlorophenyl)(6-chloro-3-pyridinyl)methanone
Inchi Code 1S/C12H7Cl2NO/c13-10-3-1-2-8(6-10)12(16)9-4-5-11(14)15-7-9/h1-7H
Inchi Key XTEHBOZQORDTAQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5625-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-15
Availability details for
Quantity
  • Availability
  • Add to Cart