RIE240870978 Rieke Metals, Inc.

5-(2-Bromobenzoyl)-2-chloropyridine

  • MDL number MFCD13153350
  • Molecular Weight : 296.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-bromophenyl)(6-chloro-3-pyridinyl)methanone
Inchi Code 1S/C12H7BrClNO/c13-10-4-2-1-3-9(10)12(16)8-5-6-11(14)15-7-8/h1-7H
Inchi Key UYNKCDKVECCFQS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5625-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-17
Availability details for
Quantity
  • Availability
  • Add to Cart