RIE240870979 Rieke Metals, Inc.

5-(3-Bromobenzoyl)-2-chloropyridine

  • MDL number MFCD13153351
  • Molecular Weight : 296.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-bromophenyl)(6-chloro-3-pyridinyl)methanone
Inchi Code 1S/C12H7BrClNO/c13-10-3-1-2-8(6-10)12(16)9-4-5-11(14)15-7-9/h1-7H
Inchi Key KEGVBAVHXNRWAQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5625-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-18
Availability details for
Quantity
  • Availability
  • Add to Cart