RIE240870980 Rieke Metals, Inc.

2-Chloro-5-(2-iodobenzoyl)pyridine

  • MDL number MFCD13153353
  • Molecular Weight : 343.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-chloro-3-pyridinyl)(2-iodophenyl)methanone
Inchi Code 1S/C12H7ClINO/c13-11-6-5-8(7-15-11)12(16)9-3-1-2-4-10(9)14/h1-7H
Inchi Key SPBDULYNMUQTTQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5625-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-20
Availability details for
Quantity
  • Availability
  • Add to Cart