RIE240870981 Rieke Metals, Inc.

2-Chloro-5-(3-iodobenzoyl)pyridine

  • MDL number MFCD13153354
  • Molecular Weight : 343.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-chloro-3-pyridinyl)(3-iodophenyl)methanone
Inchi Code 1S/C12H7ClINO/c13-11-5-4-9(7-15-11)12(16)8-2-1-3-10(14)6-8/h1-7H
Inchi Key VGRQKYJUDUJCSS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5625-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-21
Availability details for
Quantity
  • Availability
  • Add to Cart