RIE240870988 Rieke Metals, Inc.

2-Chloro-5-(4-nitrobenzoyl)pyridine

  • MDL number MFCD13153361
  • Molecular Weight : 262.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-chloro-3-pyridinyl)(4-nitrophenyl)methanone
Inchi Code 1S/C12H7ClN2O3/c13-11-6-3-9(7-14-11)12(16)8-1-4-10(5-2-8)15(17)18/h1-7H
Inchi Key CIEWPFKUAQSEAG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5625-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-31
Availability details for
Quantity
  • Availability
  • Add to Cart