RIE240870989 Rieke Metals, Inc.

2-Chloro-5-(4-isopropylbenzoyl)pyridine

  • MDL number MFCD07775856
  • Molecular Weight : 259.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5625-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-32
Availability details for
Quantity
  • Availability
  • Add to Cart