RIE240870990 Rieke Metals, Inc.

2-Chloro-5-(4-isopropoxybenzoyl)pyridine

  • MDL number MFCD13153362
  • Molecular Weight : 275.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-chloro-3-pyridinyl)(4-isopropoxyphenyl)methanone
Inchi Code 1S/C15H14ClNO2/c1-10(2)19-13-6-3-11(4-7-13)15(18)12-5-8-14(16)17-9-12/h3-10H,1-2H3
Inchi Key YXABLKYEZGHTGO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5625-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-33
Availability details for
Quantity
  • Availability
  • Add to Cart