RIE240870991 Rieke Metals, Inc.

5-(4-tert-Butylbenzoyl)-2-chloropyridine

  • MDL number MFCD07775833
  • Molecular Weight : 273.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-tert-butylphenyl)(6-chloro-3-pyridinyl)methanone
Inchi Code 1S/C16H16ClNO/c1-16(2,3)13-7-4-11(5-8-13)15(19)12-6-9-14(17)18-10-12/h4-10H,1-3H3
Inchi Key XOGOUOUUXVZUDN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5625-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-34
Availability details for
Quantity
  • Availability
  • Add to Cart