RIE240870993 Rieke Metals, Inc.

2-Chloro-5-(4-propylbenzoyl)pyridine

  • MDL number MFCD07775858
  • Molecular Weight : 259.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5625-35B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-35B
Availability details for
Quantity
  • Availability
  • Add to Cart