RIE240870994 Rieke Metals, Inc.

5-(4-Butylbenzoyl)-2-chloropyridine

  • MDL number MFCD07775832
  • Molecular Weight : 273.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5625-35C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-35C
Availability details for
Quantity
  • Availability
  • Add to Cart